Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec61a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201577.1 | NP_963871.1 | 3 - 4 | 210 | 74 | |
GenBank mRNA | AY034614.1 | - | 3 - 4 | 210 | 74 | |
BC048881.1 | - | 3 - 4 | 211 | 74 | ||
BC165352.1 | - | 2 - 3 | 176 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |