Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: per1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212439.2 | NP_997604.2 | 18 - 19 | 4521 | 91 | |
GenBank mRNA | AY597250.1 | - | 18 - 19 | 4521 | 91 | |
BC163718.1 | - | 17 - 18 | 4066 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |