Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult1st6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002599.1 | NP_001002599.1 | 1 - 2 | 500 | 105 | |
GenBank mRNA | AY937249.1 | - | 1 - 2 | 186 | 105 | |
BC075996.1 | - | 1 - 2 | 500 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |