Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: derl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024395.1 | NP_001019566.1 | 3 - 4 | 242 | 108 | |
GenBank mRNA | BC095262.1 | - | 3 - 4 | 242 | 108 | |
BC165145.1 | - | 3 - 4 | 271 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |