Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptgs2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025504.2 | NP_001020675.1 | 4 - 5 | 496 | 64 | |
GenBank mRNA | DQ494791.1 | - | 4 - 5 | 496 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |