Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045419.2 | NP_001038884.2 | 15 - 16 | 1900 | 72 | |
NM_001139438.1 | NP_001132910.1 | 14 - 15 | 2060 | 72 | ||
GenBank mRNA | BC122098.1 | - | 15 - 16 | 1895 | 72 | |
DT069701.1 | - | 6 - 7 | 661 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |