Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mylka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009304445.1 | XP_009302720.1 | 21 - 22 | 3905 | 83 | |
XM_009304446.1 | XP_009302721.1 | 21 - 22 | 3905 | 83 | ||
XM_017357739.1 | XP_017213228.1 | 21 - 22 | 3902 | 83 | ||
GenBank mRNA | AB267908.1 | - | 6 - 7 | 1262 | 83 | |
BC163913.1 | - | 6 - 7 | 1050 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |