Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001326364.1 | NP_001313293.1 | 3 - 4 | 1147 | 77 | |
NM_131557.2 | NP_571632.2 | 3 - 4 | 1473 | 77 | ||
XM_017358955.2 | XP_017214444.1 | 3 - 4 | 1208 | 77 | ||
XM_017358958.2 | XP_017214447.1 | 3 - 4 | 1281 | 77 | ||
GenBank mRNA | AF191578.1 | - | 2 - 3 | 731 | 77 | |
AY395731.1 | - | 3 - 4 | 1471 | 77 | ||
BC067710.1 | - | 3 - 4 | 1100 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |