Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131722.1 | NP_571797.1 | 2 - 3 | 495 | 101 | |
XM_005167061.4 | XP_005167118.1 | 2 - 3 | 626 | 101 | ||
GenBank mRNA | AB030897.1 | - | 2 - 3 | 495 | 101 | |
BC044128.1 | - | 2 - 3 | 330 | 101 | ||
BC059417.1 | - | 2 - 3 | 499 | 101 | ||
BC164402.1 | - | 2 - 3 | 289 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |