Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: f7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131819.2 | NP_571894.2 | 3 - 4 | 261 | 124 | |
GenBank mRNA | AY040345.1 | - | 3 - 4 | 245 | 124 | |
BC095023.1 | - | 3 - 4 | 261 | 124 | ||
BC165195.1 | - | 3 - 4 | 279 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |