Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh3b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173221.2 | NP_775328.2 | 2 - 3 | 277 | 116 | |
XM_005163981.4 | XP_005164038.1 | 2 - 3 | 321 | 116 | ||
GenBank mRNA | AF254955.1 | - | 1 - 2 | 171 | 116 | |
BC049338.1 | - | 2 - 3 | 277 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |