Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: washc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200183.1 | NP_956477.1 | 18 - 19 | 2863 | 63 | |
GenBank mRNA | BC045490.1 | - | 18 - 19 | 2863 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |