Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002211.1 | NP_001002211.1 | 2 - 3 | 329 | 68 | |
XM_005155296.4 | XP_005155353.1 | 2 - 3 | 390 | 68 | ||
XM_005155298.3 | XP_005155355.1 | 2 - 3 | 326 | 68 | ||
GenBank mRNA | BC074073.1 | - | 2 - 3 | 329 | 68 | |
BC164605.1 | - | 1 - 2 | 111 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |