Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003415.1 | NP_001003415.1 | 1 - 2 | 867 | 153 | |
GenBank mRNA | AY648747.1 | - | 1 - 2 | 867 | 153 | |
BC093108.1 | - | 1 - 2 | 868 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |