Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006054.1 | NP_001006054.1 | 5 - 6 | 1021 | 72 | |
GenBank mRNA | BC083278.1 | - | 5 - 6 | 1021 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |