Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020627.2 | NP_001018463.1 | 6 - 7 | 1170 | 89 | |
GenBank mRNA | BC095378.1 | - | 6 - 7 | 1172 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |