Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030161.2 | NP_001025332.2 | 16 - 17 | 2007 | 142 | |
XM_017358654.2 | XP_017214143.1 | 16 - 17 | 2098 | 142 | ||
GenBank mRNA | BC098603.1 | - | 16 - 17 | 2025 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |