Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slit1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034975.1 | NP_001030147.1 | 35 - 36 | 4233 | 83 | |
XM_009296502.3 | XP_009294777.1 | 35 - 36 | 4354 | 83 | ||
GenBank mRNA | BC163568.1 | - | 35 - 36 | 4221 | 83 | |
CU638802.1 | - | 35 - 36 | 4246 | 83 | ||
DQ153065.1 | - | 35 - 36 | 4233 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |