Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmpr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039807.1 | NP_001034896.1 | 4 - 5 | 1343 | 61 | |
GenBank mRNA | BC163200.1 | - | 4 - 5 | 1201 | 61 | |
DQ002899.1 | - | 4 - 5 | 1343 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |