Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077598.1 | NP_001071066.1 | 2 - 3 | 1625 | 143 | |
GenBank mRNA | BC125956.1 | - | 2 - 3 | 1625 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |