Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104937.1 | NP_001098407.1 | 13 - 14 | 1630 | 92 | |
XM_005165296.4 | XP_005165353.1 | 13 - 14 | 1698 | 92 | ||
GenBank mRNA | AM084880.1 | - | 13 - 14 | 1630 | 92 | |
AM749801.1 | - | 13 - 14 | 1630 | 92 | ||
AY324390.1 | - | 3 - 4 | 285 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |