Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131480.1 | NP_571555.1 | 9 - 10 | 1042 | 98 | |
GenBank mRNA | AJ005692.1 | - | 9 - 10 | 1042 | 98 | |
BC045863.1 | - | 9 - 10 | 1100 | 98 | ||
BC060933.1 | - | 9 - 10 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |