Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caspa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131505.2 | NP_571580.1 | 2 - 3 | 413 | 99 | |
GenBank mRNA | AF233434.1 | - | 2 - 3 | 392 | 99 | |
BC095022.1 | - | 2 - 3 | 413 | 99 | ||
BC164780.1 | - | 2 - 3 | 426 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |