Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arntl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178300.1 | NP_840085.1 | 13 - 14 | 1422 | 65 | |
XM_005166616.4 | XP_005166673.1 | 16 - 17 | 1680 | 65 | ||
XM_009303573.3 | XP_009301848.1 | 15 - 16 | 1611 | 65 | ||
XM_021477412.1 | XP_021333087.1 | 15 - 16 | 1618 | 65 | ||
GenBank mRNA | AB087257.1 | - | 13 - 14 | 1422 | 65 | |
BC134895.1 | - | 16 - 17 | 1665 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |