Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anxa1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181758.1 | NP_861423.1 | 11 - 12 | 900 | 150 | |
XM_009301449.1 | XP_009299724.1 | 11 - 12 | 956 | 150 | ||
GenBank mRNA | AY178793.1 | - | 11 - 12 | 900 | 150 | |
BC053190.1 | - | 10 - 11 | 883 | 150 | ||
BC164376.1 | - | 10 - 11 | 881 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |