Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199745.2 | NP_956039.2 | 7 - 8 | 1291 | 63 | |
GenBank mRNA | BC054666.1 | - | 927 | 63 | ||
BC153482.1 | - | 5 - 6 | 915 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |