Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199942.2 | NP_956236.1 | 3 - 4 | 592 | 101 | |
GenBank mRNA | BC057430.1 | - | 502 | 101 | ||
EH585004.1 | - | 3 - 4 | 550 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |