Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201078.1 | NP_957372.1 | 10 - 11 | 1189 | 94 | |
GenBank mRNA | BC045939.1 | - | 10 - 11 | 1189 | 94 | |
BC165833.1 | - | 10 - 11 | 1109 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |