Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcun1d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205538.1 | NP_991101.1 | 6 - 7 | 732 | 106 | |
XM_005167810.4 | XP_005167867.1 | 6 - 7 | 837 | 106 | ||
XM_005167811.4 | XP_005167868.1 | 6 - 7 | 859 | 106 | ||
XM_021478598.1 | XP_021334273.1 | 6 - 7 | 804 | 106 | ||
GenBank mRNA | BC065884.1 | - | 6 - 7 | 732 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |