Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205690.2 | NP_991253.1 | 2 - 3 | 169 | 93 | |
XM_005163003.4 | XP_005163060.1 | 3 - 4 | 252 | 93 | ||
GenBank mRNA | BC065863.1 | - | 2 - 3 | 173 | 93 | |
EU348854.1 | - | 1 - 2 | 139 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |