Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rlbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205690.2 | NP_991253.1 | 5 - 6 | 711 | 81 | |
XM_005163003.4 | XP_005163060.1 | 6 - 7 | 794 | 81 | ||
GenBank mRNA | BC065863.1 | - | 5 - 6 | 715 | 81 | |
EU348854.1 | - | 4 - 5 | 681 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |