Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212876.1 | NP_998041.1 | 1 - 2 | 329 | 96 | |
GenBank mRNA | BC066748.1 | - | 1 - 2 | 329 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |