Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asic4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214786.1 | NP_999951.1 | 5 - 6 | 1385 | 68 | |
GenBank mRNA | AJ609620.1 | - | 5 - 6 | 1385 | 68 | |
BC095568.1 | - | 5 - 6 | 1392 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |