Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002160.2 | NP_001002160.2 | 5 - 6 | 768 | 126 | |
GenBank mRNA | BC071307.1 | - | 5 - 6 | 768 | 126 | |
EH517711.1 | - | 3 - 4 | 762 | 126 | ||
EH605435.1 | - | 3 - 4 | 757 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |