Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nutf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003598.2 | NP_001003598.1 | 3 - 4 | 393 | 66 | |
GenBank mRNA | BC078197.1 | - | 3 - 4 | 302 | 66 | |
EH475711.1 | - | 2 - 3 | 433 | 66 | ||
EH538050.1 | - | 3 - 4 | 431 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |