Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osbpl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004646.1 | NP_001004646.1 | 20 - 21 | 2304 | 59 | |
XM_005170028.4 | XP_005170085.1 | 20 - 21 | 2558 | 59 | ||
GenBank mRNA | BC078429.1 | - | 20 - 21 | 2304 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |