Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: il12ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007108.1 | NP_001007109.1 | 7 - 8 | 983 | 86 | |
GenBank mRNA | AB183002.1 | - | 7 - 8 | 983 | 86 | |
BC121764.1 | - | 7 - 8 | 982 | 86 | ||
BC164577.1 | - | 6 - 7 | 954 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |