Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nabp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008643.1 | NP_001008643.1 | 3 - 4 | 523 | 87 | |
GenBank mRNA | BC086713.1 | - | 3 - 4 | 523 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |