Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017727.1 | NP_001017727.1 | 5 - 6 | 642 | 131 | |
XM_005169364.4 | XP_005169421.1 | 5 - 6 | 612 | 131 | ||
XM_005169365.4 | XP_005169422.1 | 5 - 6 | 649 | 131 | ||
GenBank mRNA | BC093230.1 | - | 5 - 6 | 642 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |