Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033106.1 | NP_001028278.1 | 3 - 4 | 308 | 74 | |
GenBank mRNA | BC100049.1 | - | 3 - 4 | 308 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |