Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacna2d1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044960.2 | NP_001038425.2 | 37 - 38 | 3182 | 63 | |
XM_005164746.4 | XP_005164803.1 | 37 - 38 | 3394 | 63 | ||
XM_009300287.3 | XP_009298562.1 | 38 - 39 | 3418 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Danio rerio Probe spans exons |