Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gas2l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045098.1 | NP_001038563.1 | 7 - 8 | 823 | 73 | |
XM_005164456.4 | XP_005164513.2 | 7 - 8 | 880 | 73 | ||
XM_021475058.1 | XP_021330733.1 | 7 - 8 | 791 | 73 | ||
GenBank mRNA | BC134975.1 | - | 7 - 8 | 766 | 73 | |
JX013520.1 | - | 6 - 7 | 654 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |