Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131220.1 | NP_571295.1 | 4 - 5 | 405 | 72 | |
XM_005160619.4 | XP_005160676.1 | 4 - 5 | 459 | 72 | ||
XM_005160620.4 | XP_005160677.1 | 3 - 4 | 431 | 72 | ||
XM_005160621.4 | XP_005160678.1 | 3 - 4 | 417 | 72 | ||
GenBank mRNA | BC044453.1 | - | 3 - 4 | 367 | 72 | |
BC059187.1 | - | 3 - 4 | 1820 | 72 | ||
BC066760.1 | - | 4 - 5 | 405 | 72 | ||
BC164908.1 | - | 3 - 4 | 319 | 72 | ||
L11711.1 | - | 3 - 4 | 380 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |