Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: u2af1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177479.2 | NP_803432.1 | 1 - 2 | 42 | 73 | |
GenBank mRNA | AF506202.1 | - | 1 - 2 | 42 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons |