Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glud1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199545.4 | NP_955839.2 | 1 - 2 | 608 | 64 | |
GenBank mRNA | AY577003.1 | - | 1 - 2 | 608 | 64 | |
BC044202.1 | - | 1 - 2 | 544 | 64 | ||
BC063940.1 | - | 1 - 2 | 472 | 64 | ||
BC165763.1 | - | 1 - 2 | 426 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |