Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serhl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199615.2 | NP_955909.2 | 7 - 8 | 577 | 101 | |
GenBank mRNA | BC044494.1 | - | 7 - 8 | 577 | 101 | |
BC066637.1 | - | 7 - 8 | 510 | 101 | ||
EH457337.1 | - | 7 - 8 | 559 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |