Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mocs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200127.1 | NP_956421.1 | 6 - 7 | 781 | 64 | |
XM_021467123.1 | XP_021322798.1 | 3 - 4 | 270 | 64 | ||
GenBank mRNA | BC042324.1 | - | 6 - 7 | 781 | 64 | |
BC071459.1 | - | 6 - 7 | 743 | 64 | ||
BC164541.1 | - | 6 - 7 | 663 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |