Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201456.1 | NP_958864.1 | 11 - 12 | 1204 | 111 | |
GenBank mRNA | AY398349.1 | - | 11 - 12 | 1204 | 111 | |
BC049425.1 | - | 10 - 11 | 985 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |