Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205729.2 | NP_991292.2 | 5 - 6 | 1606 | 77 | |
GenBank mRNA | AY391444.1 | - | 5 - 6 | 1407 | 76 | |
BC095127.1 | - | 5 - 6 | 1575 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |