Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnase1l1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002403.1 | NP_001002403.1 | 5 - 6 | 503 | 69 | |
GenBank mRNA | BC076179.1 | - | 5 - 6 | 503 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |